Learn More
Impacto™ Anti-Impact Gloves with Wrist Support
Impacto™ 704-20 Glove with padding in the palm and thumb web area protects from impact and vibration and wrist support protects from RSI
$45.60
Specifications
Material Leather, Nylon Lycra
View More Specs
Products 3
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-031939
View Documents
Impacto™
AV70420PMED
Medium
Pair for $45.60
Add to cart
19-031943
View Documents
Impacto™
AV70420PXLG
X-Large
Pair for $45.60
Add to cart
19-031936
View Documents
Impacto™
AV70420PLRG
Large This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
Impacto™ Anti-Impact Glove with Wrist Support
  • Fingerless nylon Lycra and grain leather glove
  • VEP padding in the palm and thumb web area
  • Detachable grain leather wrist support minimizes extension and over flexion of the wrist to prevent RSI such as carpal tunnel syndrome or tendinitis
  • Provides protection from abrasion, impact and vibration
  • Fingerless style for optimum dexterity
  • Used with hand or electric tools in repetitive tasks
Specifications
Specifications
Leather, Nylon Lycra