Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Impacto™ Fingerless Anti-Impact Gloves with Detachable Wrist Support
Click to view available options
Size:
Medium
Large
X-Large
Hand:
Left
Specific
Description
- VEP padding in the palm and thumb web area
- Detachable grain leather wrist support
- Fingerless style for optimum dexterity
- Protection from abrasion, impact and shock
- Minimizes extension and over flexion of the wrist to prevent RSI
- Use with hand or electric tools in repetitive tasks
Specifications
Back Material | Spandex |
Cuff Material | Grain Leather |
Color | Blue, Yellow |
Hand | Specific |
Material | Grain Leather |
Product Type | Anti-impact Gloves |
Size | Large |
Finger Style | Fingerless |
For Use With (Application) | Construction, Assembly, Manufacturing, Maintenance, and Warehousing |
Glove Cuff Color | Yellow |
Show More |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction