Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Impacto™ Fingerless Nylon Gloves with VEP Padding

Gloves are constructed of nylon and pearl leather with VEP padding that extends from the palm to the wrist joint for impact and shock protection.

$14.71

Specifications

Color Blue
For Use With (Application) General Assembly and Material Handling
Material Nylon Lycra™
Product Type Anti-Impact Gloves
Series 501-35
View More Specs

Products 8
Catalog Number Mfr. No. Size Hand Price Quantity & Availability  
Catalog Number Mfr. No. Size Hand Price Quantity & Availability  
18-999-4604A
View Documents
Impacto™
50135110021
Small Left
Each for $14.71
Add to cart
 
18-999-4603A
View Documents
Impacto™
50135110022
Small Right
Each for $14.71
Add to cart
 
18-999-4604B
View Documents
Impacto™
50135110031
Medium Left
Each for $14.71
Add to cart
 
18-999-4603B
View Documents
Impacto™
50135110032
Medium Right
Each for $14.71
Add to cart
 
18-999-4604C
View Documents
Impacto™
50135110041
Large Left
Each for $14.71
Add to cart
 
18-999-4603C
View Documents
Impacto™
50135110042
Large Right
Each for $14.71
Add to cart
 
18-999-4604D
View Documents
Impacto™
50135110051
X-Large Left
Each for $14.71
Add to cart
 
18-999-4603D
View Documents
Impacto™
50135110052
X-Large Right
Each for $14.71
Add to cart
 
Description

Description

  • Stretch Lycra™ outer fabric for comfort, dexterity, and moisture evaporation
  • Visco elastic polymer (VEP) padding absorbs impact, and dissipates shock
  • Ideal for general assembly, and material handling
Specifications

Specifications

Blue
Nylon Lycra™
501-35
General Assembly and Material Handling
Anti-Impact Gloves
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit