Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Impacto™ Anti-Impact Glove Liners with Extended VEP Padding

Click to view available options
Size:
Small
Medium
Large
X-Large
Hand:
Left
Right
This item is not returnable. View return policy
This item is not returnable. View return policy

Stretch polycotton fabric ensures optimum breathability and comfort. VEP padding that extends from the palm to the wrist joint provides users with additional protection.

  • 4-way stretch polycotton fabric ensures optimum breathability and comfort
  • Contoured VEP 1/8 in. padding in the palm extends to the wrist joint for additional protection
  • Open-finger design for tactile feedback and mobility
  • Designed to be worn under work gloves
  • Offers protection from electric, manual, and power tools
  • Use with applications such as construction, assembly, manufacturing, maintenance, and warehousing
Color Blue
Hand Right
Material Polycotton
Product Type Glove Liners
Size Medium
Finger Style Fingerless
For Use With (Application) Construction, Assembly, Manufacturing, Maintenance, and Warehousing
Glove Cuff Style Open
Series 510-00
Length (English) 8 in.
Length (Metric) 20.5 cm
Show More Show Less