Learn More
Impacto™ Full Finger Anti-Impact Glove Liner
For extended use with heavy-impact tools
Manufacturer:  Impacto™ 60100PL
 View more versions of this product
Catalog No. 19-048-714C
Add to cart
Description
Description
  • Full finger polycotton lycra glove liner
  • Impact absorbing VEP padding in the palm and fingers
  • Recommended to be worn under work glove or by itself in clean applications
  • Provides impact and shock protection
  • Soft polycotton ensures breathability and hand dexterity
Specifications
Specifications
Safety
23 to 25cm
Soft
Lycra™
9 in.
9 to 10 in.
Full
Visco Elastic Polymer padding in the palm, finger
Large
23 cm