Learn More
Impacto™ Anti-Impact Glove Liner Full Finger
Impacto™ 601-00 Anti-Impact Glove Liner to wear inside work glove padded in the palm and fingers
$35.40
Specifications
Material Polycotton
Exterior Finish Soft
Finger Style Full
Includes Visco Elastic Polymer padding in the palm, finger
View More Specs
Products 3
Catalog Number Mfr. No. Circumference (English) Hand Length (English) Size Price Quantity & Availability  
Catalog Number Mfr. No. Circumference (English) Hand Length (English) Size Price Quantity & Availability  
19-048-714C
View Documents
Impacto™
60100PL
9 to 10 in. 9 in. Large Pair for $35.40
Add to cart
19-048-714B
View Documents
Impacto™
60100PM
8 to 9 in. 8 in. Medium Pair for $35.40
Add to cart
19-048-714D
View Documents
Impacto™
60100PXL
10 to 11 in. 11 in. X-Large Pair for $35.40
Add to cart
Description
Description
Impacto™ Anti-Impact Glove Liners
  • Full finger made with polycotton lycra
  • VEP padding in the palm and fingers
  • Recommended to be worn under work glove or by itself in clean applications
  • Provides impact and vibration protection
  • Soft polycotton ensures breathability and hand dexterity
Specifications
Specifications
Polycotton
Soft
Full
Visco Elastic Polymer padding in the palm, finger