Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Impacto™ Fingerless Leather Palm Tool Grip Gloves

Catalog Number 19031902
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
Hand:
Left
Right
Specific
This item is not returnable. View return policy
This item is not returnable. View return policy

Nylon gloves are constructed with contoured VEP padding to provide protection from vibration, impact, and abrasion. The grain leather palm is smooth and provides extended wear.

  • Protection from vibration, impact, and abrasion
  • Grain leather palm is soft and durable
  • Fingerless style ensures tactile feedback
  • Use for protection from hand tools and power tools
Back Material Nylon Lycra™
Color Blue, Yellow
Hand Specific
Material Grain Leather
Product Type Tool Gloves
Size Medium
Finger Style Fingerless
For Use With (Application) Protection From Hand Tools and Power Tools
Glove Cuff Style Open
Series 501-20
Length (English) 8 in.
Length (Metric) 20.5 cm
Show More Show Less
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit