Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Impacto™ Fingerless Anti-Impact Gloves with Suede Leather Palms/Sides

Catalog Number 19031980
Click to view available options
Size:
Small
X-Large
Hand:
Left
Right
This item is not returnable. View return policy
This item is not returnable. View return policy

VEP padding throughout the glove offers ideal protection during automotive applications, while the suede leather cover provides abrasion protection.

  • Nylon fabric ensures breathability and comfort
  • Contoured VEP 1/8 in. padding in the palm, edge, and back of the hand provides protection from impact and shock
  • Fingerless design for tactile feedback and mobility
  • Suede leather cover offers abrasion protection
  • Side padding protects from impact during automotive applications
Back Material Nylon
Color Blue, Gray
Hand Left
Material Suede
Product Type Anti-impact Gloves
Size X-Large
Finger Style Fingerless
For Use With (Application) Automotive Assembly
Glove Cuff Style Open
Series 503-10
Length (English) 11 in.
Length (Metric) 28 cm
Show More Show Less
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit