Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IMPDH1 Rabbit anti-Human, Polyclonal, Bioss
Rabbit Polyclonal Antibody
Manufacturer: Bioss BS6256R
Description
The protein encoded by this gene acts as a homotetramer to regulate cell growth. The encoded protein is an enzyme that catalyzes the synthesis of xanthine monophosphate (XMP) from inosine-5 ft.-monophosphate (IMP). This is the rate-limiting step in the de novo synthesis of guanine nucleotides. Defects in this gene are a cause of retinitis pigmentosa type 10 (RP10). Several transcript variants encoding different isoforms have been found for this gene.Specifications
IMPDH1 | |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
IMPDH1 | |
Liquid | |
Rabbit | |
IgG | |
100 μL | |
-20°C | |
Primary | |
3614 |
IMPDH1 | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
P20839 | |
B930086D20Rik; BOS_4522; DKFZp781N0678; HAMAP-Rule:MF_03156}; IMP (inosine 5′-monophosphate) dehydrogenase 1; IMP (inosine monophosphate) dehydrogenase 1; IMP dehydrogenase 1; IMP dehydrogenase 1 {ECO:0000255; IMPD; IMPD 1; IMPD 1 {ECO:0000255; IMPD1; IMPDH 1; IMPDH 1 {ECO:0000255; Impdh1; IMPDH-I; inosine 5′-phosphate dehydrogenase 1; inosine monophosphate dehydrogenase 1; Inosine-5′-monophosphate dehydrogenase 1; inosine-5′-monophosphate dehydrogenase 1 {ECO:0000255; LCA11; RP10; sWSS2608 | |
KLH conjugated synthetic peptide derived from human IMPDH1. | |
Protein A | |
RUO | |
Antibody | |
Polyclonal | |
Human |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title