Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Impentamine dihydrobromide, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Potent and highly selective histamine H3 receptor antagonist (pA2 = 8.4); displays > 30000-fold selectivity over H1 and H2 receptors. Can act as a partial agonist in SK-N-MC cells expressing human H3 receptors. Produces antinociception, possibly via a non-H1, -H2 or -H3 receptor-mediated mechanism, following central administration in vivo.
Specifications
Quantity | 10 mg |
Formula Weight | 315.05 |
Percent Purity | >98% |
Chemical Name or Material | Impentamine dihydrobromide |
Safety and Handling
Recommended Storage | Desiccate at Room Temperature |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction