Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Impentamine dihydrobromide, Tocris Bioscience™

Click to view available options
Quantity:
10mg
50mg
This item is not returnable. View return policy
149629-70-9
C8H17Br2N3
315.053
QSDXAAWBBPWGEL-UHFFFAOYSA-N
impentamine dihydrobromide,4-5-aminopentyl imidazole dihydrobromide,5-1h-imidazol-4-yl pentan-1-amine dihydrobromide,5-1h-imidazol-5-yl pentan-1-amine;dihydrobromide,5-1h-imidazol-5-yl pentan-1-amine-hydrogen bromide 1/2
53439849
5-(1H-imidazol-5-yl)pentan-1-amine;dihydrobromide
C1=C(NC=N1)CCCCCN.Br.Br
This item is not returnable. View return policy
149629-70-9
C8H17Br2N3
315.053
QSDXAAWBBPWGEL-UHFFFAOYSA-N
impentamine dihydrobromide,4-5-aminopentyl imidazole dihydrobromide,5-1h-imidazol-4-yl pentan-1-amine dihydrobromide,5-1h-imidazol-5-yl pentan-1-amine;dihydrobromide,5-1h-imidazol-5-yl pentan-1-amine-hydrogen bromide 1/2
53439849
5-(1H-imidazol-5-yl)pentan-1-amine;dihydrobromide
C1=C(NC=N1)CCCCCN.Br.Br

Selective H3 antagonist

Potent and highly selective histamine H3 receptor antagonist (pA2 = 8.4); displays > 30000-fold selectivity over H1 and H2 receptors. Can act as a partial agonist in SK-N-MC cells expressing human H3 receptors. Produces antinociception, possibly via a non-H1, -H2 or -H3 receptor-mediated mechanism, following central administration in vivo.
Quantity 50mg
Formula Weight 315.05
Percent Purity >98%
Chemical Name or Material Impentamine dihydrobromide

Recommended Storage : Desiccate at Room Temperature