Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IMPG1 Rabbit anti-Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen IMPG1
Applications Immunohistochemistry, Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB094498
View Documents
Novus Biologicals
NBP304853100UL
100 μg Each for $320.71
Add to cart
 
NB094499
View Documents
Novus Biologicals
NBP30485320UL
20 μg Each for $138.37
Add to cart
 
Description

Description

IMPG1 Polyclonal antibody specifically detects IMPG1 in Rat samples. It is validated for Western Blot, Immunohistochemistry
Specifications

Specifications

IMPG1
Unconjugated
Rabbit
Rat
GP147, interphotoreceptor matrix proteoglycan 1, IPM150, SPACR
Recombinant fusion protein containing a sequence corresponding to amino acids 21-300 of human IMPG1 (NP_001554.2).
Polyclonal
Affinity purified
Immunohistochemistry, Western Blot
Purified
RUO
PBS with 50% glycerol, pH7.3.
3617
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit