Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Importin 11 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP303801100UL
Description
Importin 11 Polyclonal specifically detects Importin 11 in Human, Mouse samples. It is validated for Western Blot.Specifications
Importin 11 | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
51194 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot | |
Unconjugated | |
imp11, importin 11, Ran binding protein 11, Ran-binding protein 11, RANBP11, RanBP11importin-11, SLRN | |
Recombinant fusion protein containing a sequence corresponding to amino acids 776-975 of human Importin 11 (NP_057422.3). | |
100 μL | |
Primary | |
Human, Mouse | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction