Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Importin 13 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA530326
Description
Recommended positive controls: human IPO13 transfected 293T. Predicted reactivity: Mouse (98%), Rat (98%), Chicken (82%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a member of the importin-beta family of nuclear transport proteins. The encoded protein mediates the import of specific cargo proteins from the cytoplasm to the nucleus and is dependent on the Ras-related nuclear protein-GTPase system. The encoded protein is also involved in nuclear export of the eukaryotic translation initiation factor 1A.
Specifications
Importin 13 | |
Polyclonal | |
Unconjugated | |
IPO13 | |
Imp13; importin 13; importin-13; Ipo13; Kap13; karyopherin 13; karyopherin-13; KIAA0724; late gestation lung 2; late gestation lung 2 protein; LGL2; Ran binding protein 13; ran-binding protein 13; RanBP13; RP11-7O11.4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
9670 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
O94829 | |
IPO13 | |
Recombinant fragment corresponding to a region within amino acids 93 and 310 of Human Importin 13. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction