Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Importin-9 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen Importin-9
Applications Immunofluorescence, Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB093163
View Documents
Novus Biologicals
NBP303818100UL
100 μg
Each for $320.71
Add to cart
 
NB093164
View Documents
Novus Biologicals
NBP30381820UL
20 μg
Each for $138.37
Add to cart
 
Description

Description

Importin-9 Polyclonal antibody specifically detects Importin-9 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence
Specifications

Specifications

Importin-9
Unconjugated
Rabbit
Core ESC Like Genes, Stem Cell Markers
PBS with 50% glycerol, pH7.3.
55705
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Immunofluorescence, Western Blot
Purified
RUO
Human, Mouse, Rat
DKFZp761M1547, FLJ10402, Imp9IMP9, importin 9, importin-9, KIAA1192, Ran-binding protein 9, RANBP9
Recombinant fusion protein containing a sequence corresponding to amino acids 827-1041 of human Importin-9 (NP_060555.2).
Polyclonal
Affinity purified
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit