Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Importin beta-1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5115424
Description
Antibody detects endogenous levels of total Importinbeta.
Nucleocytoplasmic transport, a signal- and energy-dependent process, takes place through nuclear pore complexes embedded in the nuclear envelope. The import of proteins containing a nuclear localization signal requires the NLS import receptor, a heterodimer of importin alpha and beta subunits also known as karyopherins. Importin alpha binds the NLS-containing cargo in the cytoplasm and importin beta docks the complex at the cytoplasmic side of the nuclear pore complex. In the presence of nucleoside triphosphates and the small GTP binding protein Ran, the complex moves into the nuclear pore complex and the importin subunits dissociate. Importin alpha enters the nucleoplasm with its passenger protein and importin beta remains at the pore. Interactions between importin beta and the FG repeats of nucleoporins are essential in translocation through the pore complex. The protein encoded by this gene is a member of the importin beta family.
Specifications
Importin beta-1 | |
Polyclonal | |
Unconjugated | |
KPNB1 | |
AA409963; HGNC:6400; IMB1; Impnb; importin 1; importin 90; importin beta; importin beta-1 subunit; importin subunit beta-1; Importin-90; IPO1; IPOB; KapB1; karyopherin (importin) beta 1; karyopherin beta 1; karyopherin subunit beta 1; karyopherin subunit beta-1; karyopherin,beta 1; Kpnb1; MGC2155; MGC2156; MGC2157; NTF97; Nuclear factor p97; pore targeting complex 97 kDa subunit; PTAC97; SCG | |
Rabbit | |
Affinity Chromatography | |
RUO | |
16211, 24917, 3837 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide | |
P52296, P70168, Q14974 | |
KPNB1 | |
A synthesized peptide derived from human KPNB1(Accession Q14974), corresponding to amino acid residues D756-G806. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction