Learn More
INA Mouse anti-Mouse, Clone: 257CT7.1.2, Abnova™
Mouse monoclonal antibody raised against synthetic peptide of mouse INA.
Manufacturer:  Abnova Corporation MAB12313
Catalog No. 89-934-376
Add to cart
Description
Description
Neurofilaments are type IV intermediate filament heteropolymers composed of light, medium, and heavy chains. Neurofilaments comprise the axoskeleton and they functionally maintain the neuronal caliber. They may also play a role in intracellular transport to axons and dendrites. This gene is a member of the intermediate filament family and is involved in the morphogenesis of neurons. [provided by RefSeq]
Specifications
Specifications
INA
257CT7.1.2
Mouse monoclonal antibody raised against synthetic peptide of mouse INA.
In PBS (0.09% sodium azide)
FLJ18662/FLJ57501/MGC12702/NEF5/NF-66/TXBP-1
Mouse
IgG1, kappa
400 uL
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Mouse
Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
Immunohistochemistry (1:10-1:50) Immunofluorescence (1:10-1:50) Western Blot (1:100-1:250) The optimal working dilution should be determined by the end user.
Liquid
INA
A synthetic peptide (conjugated with KLH) corresponding to amino acids 290-319 of human INA.
Protein G purification
RUO
Primary
9118