Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

INCA1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548921

Catalog No. PA548921

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

INCA1
Polyclonal
PBS with 0.09% sodium azide
Q0VD86, Q6PKN7
Protein INCA1, Inhibitor of CDK interacting with cyclin A1, INCA1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
103844, 388324
Western Blot
Unconjugated
INCA1
Liquid
INCA1
KLH conjugated synthetic peptide between 14-42 amino acids from the N-terminal region of human INCA1.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit