Learn More
Azzota Corp INCB-24360 DFS Item
Manufacturer:  Azzota Corp C1180
INCB-024360 (Epacadostat) is an orally available inhibitor of indoleamine 2,3-dioxygenase (IDO1), with potential immunomodulating and antineoplastic activities. INCB-024360 targets and binds to IDO1. Please request if need more quantities
Catalog No. 50-161-3129