Learn More
Selleck Chemical LLCSupplier Diversity Partner INCB053914 DFS Item
Manufacturer:  Selleck Chemical LLCSupplier Diversity Partner S88001MG
INCB053914 is a novel, ATP-competitive, small molecule, pan-inhibitor of PIM kinases with IC50 values of 0.24 nM, 30 nM and 0.12 nM for PIM1, PIM2 and PIM3 respectively in biochemical assays.
Catalog No. 50-194-8384