Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Inconel™ 600 gauze, 20 mesh woven from 0.41mm (0.016 in.) dia. wire

$15.03 - $99.20

Specifications

Chemical Name or Material Inconel™ 600 gauze
Name Note 20 mesh woven from 0.41mm (0.016 in.) dia. wire
MDL Number MFCD01741927
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AA46510FL View Documents Alfa Aesar™
46510FL
75 x 75 mm Each for $15.03
Order in Bulk

Request bulk or custom formats

Add to cart
 
AA46510HB View Documents Alfa Aesar™
46510HB
300 x 300 mm Each for $99.20
Order in Bulk

Request bulk or custom formats

Add to cart
 
AA46510GT View Documents Alfa Aesar™
46510GT
150 x 150 mm Each for $26.90
Order in Bulk

Request bulk or custom formats

Add to cart
 
Specifications

Specifications

Inconel™ 600 gauze
20 mesh woven from 0.41mm (0.016 in.) dia. wire
MFCD01741927
Safety and Handling

Safety and Handling

GHS H Statement
H372-H351-H317
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing cancer.
May cause an allergic skin reaction.

P201-P202-P260-P264b-P270-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c

H317-H350-H372

TSCA : Yes

Recommended Storage : Ambient temperatures

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit