Alfa Aesar™ Inconel™ 600 gauze, 200 mesh woven from 0.05mm (0.002 in.) dia. wire
Manufacturer: Alfa Aesar™ 46791GT
Quantity 150 x 150mm
 View more versions of this product
Catalog No. AA46791GT
  Check Availability
Add to Cart
Related Products
Product Content Feedback