Learn More
Inconel™ 600 gauze, 40 mesh woven from 0.25mm (0.01 in.) dia. wire, Thermo Scientific Chemicals
$38.35 - $123.90
Specifications
Chemical Name or Material | Inconel™ 600 gauze |
---|---|
Name Note | 40 mesh woven from 0.25mm (0.01 in.) dia. wire |
Avg. Mol. Wt. or Mol. Wt. Range | Ni:Cr:Fe; 77:15.5:7.5 wt% |
MDL Number | MFCD01741927 |
Solubility Information | Insoluble in water. |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
AA46745HB
|
Thermo Scientific Chemicals
046745HB |
300 x 300 mm |
Each for $123.90
Order in Bulk
Request bulk or custom formats |
|
|||||
AA46745GT
|
Thermo Scientific Chemicals
046745GT |
150 x 150 mm |
Each for $38.35
Order in Bulk
Request bulk or custom formats |
|
|||||
Description
Applications include heaters, stills, bubble towers and condensers for processing of fatty acids; evaporator tubes, tube sheets and flaking trays for the manufacture of sodium sulfide; and equipment for handling abietic acid in the manufacture of paper pulp. It is used for retorts, muffles, roller hearths and other furnace components and for heat-treating baskets and trays. The alloy is a standard material of construction for nuclear reactors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Inconel™ 600 gauze | |
Ni:Cr:Fe; 77:15.5:7.5 wt% | |
Insoluble in water. | |
Wire Cloth |
40 mesh woven from 0.25mm (0.01 in.) dia. wire | |
MFCD01741927 | |
40 Mesh |
Safety and Handling
GHS H Statement
H372-H351-H317
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing cancer.
May cause an allergic skin reaction.
P201-P202-P260-P264b-P270-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
H317-H350-H372
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.