Learn More
Chemglass Life Sciences Incubator, 10L, Incu-Shake, with Non-Slip Rubber Mat, 230V
SDP
Supplier:  Chemglass Life Sciences CLS254302E
Encompass_Preferred
Incubator, 10L, Incu-Shake, with Non-Slip Rubber Mat, 230V. The new Incu-Shaker™ series of shaking incubators provide digitally regulated temperature with precisely controlled circular (orbital) mixing, optimized for the culture of a wide variety of cell types.The 10L Incu-Shakers™ are equipped with a large 18 x 18 inch platform for high capacity applications.The standard platform (included with both models) features a non-slip, rubber coated surface, ideal for tissue culture flasks, Petri dishes and staining trays.Temperature, shaking speed and run time are displayed on the large LED control panel located on the front of the unit. The integral microprocessor includes a constant monitoring system that verifies and maintains accuracy through the duration of the program. Sophisticated over-temperature and over-speed controls ensure long life, safety and sample integrity.
Catalog No. 50-143-361
Explore available promotions
Add to Cart
Request a Quote
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.