Thermo Scientific™ Cell Roller System Accessories for Environmental Chambers and Incubators
Allow production of monolayer cell cultures in standard roller culture vessels.
Manufacturer: Thermo Scientific™ 228077
 View more versions of this product
Catalog No. 22-807-7
  Check Availability
Add to Cart
    Specifications
    For Use With (Equipment) Cell Roller System (4868)
    Product Type Cell Roller System
    Installation Factory installed
    Description Alarm system for Cell Rollers
    Allow production of monolayer cell cultures in standard roller culture vessels with these Cell Roller System Accessories for Thermo Scientific™ Environmental Chambers and Incubators. Oxygenation and exposure of the cells to the media growth area are improved. Culture yields are increased by the uniform temperature control and cell roll system’s continuous, gentle rotation.
  • Description & Specifications
Related Products
Product Content Feedback