Learn More
Chemglass Life SciencesSupplier Diversity Partner Incubator, Refrigerated, Shaking, Orbital, 120V DFS Item
Manufacturer:  Chemglass Life SciencesSupplier Diversity Partner CLS2444R06
Incubator, Refrigerated, Shaking, Orbital, 120V. This series of orbital shaking incubators feature a unique adjustable counter balance weighting system to accommodate off-center loads, varying capacities & stroke lengths, allowing for smoother operation. An adjustable orbit allows the user to change the rotational stroke from 0.5 inches (vigorous shaking) to 1 inch (gentle shaking) to accommodate different types of cells & provide maximum oxygen transfer. Units have small footprints & their chambers include a shelf. The –R06 & -RH6 models refrigerate via a 1/6 horsepower motorized compressor (produced without CFCs/HCFCs).All major functions: temperature, RPM & time have alarms that alert the user to deviations from set parameters. Over temperature protection is provided by a safety thermostat. Fluorescent light in the unit provides clear visibility of the chamber interior. Access port included for recording & monitoring accessories. Supplied with one platform & one shelf.
Catalog No. 50-143-529
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.