Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Indane-1-acetic acid, 97%

Click to view available options
Quantity:
1g
5g
38425-65-9
C11H12O2
176.215
MFCD00156939
RJVZEPWRJJBXLH-UHFFFAOYSA-N
1-indanylacetic acid,1-indanacetic acid,2,3-dihydro-1h-inden-1-ylacetic acid,2-2,3-dihydro-1h-inden-1-yl acetic acid,1h-indene-1-acetic acid, 2,3-dihydro,2,3-dihydro-1h-indene-1-acetic acid,1h-indene-1-aceticacid, 2,3-dihydro,1h-indene-1-acetic acid,2,3-dihydro,1-indanacetic acid 7ci,indanylacetic acid
96501
2-(2,3-dihydro-1H-inden-1-yl)acetic acid
C1CC2=CC=CC=C2C1CC(=O)O
38425-65-9
C11H12O2
176.215
MFCD00156939
RJVZEPWRJJBXLH-UHFFFAOYSA-N
1-indanylacetic acid,1-indanacetic acid,2,3-dihydro-1h-inden-1-ylacetic acid,2-2,3-dihydro-1h-inden-1-yl acetic acid,1h-indene-1-acetic acid, 2,3-dihydro,2,3-dihydro-1h-indene-1-acetic acid,1h-indene-1-aceticacid, 2,3-dihydro,1h-indene-1-acetic acid,2,3-dihydro,1-indanacetic acid 7ci,indanylacetic acid
96501
2-(2,3-dihydro-1H-inden-1-yl)acetic acid
C1CC2=CC=CC=C2C1CC(=O)O
Quantity 5g
Beilstein 1949543
Formula Weight 176.21
Physical Form Solid
Percent Purity 97%
Chemical Name or Material Indane-1-acetic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : NK8225700