Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Indapamide, USP, 98-101%, Spectrum™

Click to view available options
Quantity:
1 kg
Packaging:
Amber Glass Bottle

I1064, 26807-65-8, C16H16ClN3O3S

Spectrum™ Indapamide, USP is used to treat high blood pressure and to reduce extra salt and fluid in the body (edema). All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Amber Glass Bottle
Quantity 1 kg
Ignition Residue 0.001
Loss on Drying 0.03
CAS 26807-65-8
CAS Max % 1
Molecular Formula C16H16ClN3O3S
InChI Key NDDAHWYSQHTHNT-UHFFFAOYNA-N
SMILES CC1CC2=CC=CC=C2N1NC(=O)C1=CC=C(Cl)C(=C1)S(N)(=O)=O
IUPAC Name 4-chloro-N-(2-methyl-2,3-dihydro-1H-indol-1-yl)-3-sulfamoylbenzamide
Molecular Weight (g/mol) 365.83
Percent Purity 98 to 101%
Show More Show Less