Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Indazole-6-boronic acid pinacol ester, 97%, Acros Organics™

Click to view available options
Quantity:
1g
5g
861905-87-5
C13H17BN2O2
244.1
861905-87-5
C13H17BN2O2
244.1
Packaging Glass Bottle
Infrared Spectrum Authentic
Quantity 1g
Melting Point 134.0°C to 139.0°C
NMR Consistent with structure (1H−NMR)
Assay Percent Range 96% min. (GC)
Formula Weight 244.1
Percent Purity 97%
Chemical Name or Material Indazole-6-boronic acid pinacol ester

GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell.

GHS Signal Word: Warning