Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Indian Hedgehog/Ihh Antibody (9L16), Alexa Fluor™ 532, Novus Biologicals™
Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP222020AF532
Description
Indian Hedgehog/Ihh Monoclonal specifically detects Indian Hedgehog/Ihh in Human samples. It is validated for Western Blot, ELISA.Specifications
Indian Hedgehog/Ihh | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
IHH | |
Human recombinant Indian Hedgehog/Ihh (N-terminus) | |
Primary | |
Human | |
IgG2 |
Western Blot, ELISA | |
9L16 | |
Western Blot, ELISA | |
BDA1Indian hedgehog homolog (Drosophila), HHG2, HHG-2, Indian hedgehog, Indian hedgehog (Drosophila) homolog, Indian hedgehog homolog, indian hedgehog protein | |
Rat | |
0.1 mL | |
3549 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction