Learn More
Indian Hedgehog/Ihh Rat anti-Human, Alexa Fluor™ 594, Clone: 9L16, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP222020AF594
Catalog No. NP222020A94
Add to cart
Description
Description
Indian Hedgehog/Ihh Monoclonal antibody specifically detects Indian Hedgehog/Ihh in Human samples. It is validated for Western Blot,ELISA.
Specifications
Specifications
Indian Hedgehog/Ihh
9L16
Western Blot, ELISA
BDA1Indian hedgehog homolog (Drosophila), HHG2, HHG-2, Indian hedgehog, Indian hedgehog (Drosophila) homolog, Indian hedgehog homolog, indian hedgehog protein
Rat
IgG2
Store at 4°C in the dark.
Monoclonal
Human
ELISA, Western Blot
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
IHH
This antibody was produced from a hybridoma (rat myeloma fused with spleen cells from a rat immunized with purified recombinant human Indian Hedgehog Nterminus. Rat anti human Indian Hedgehog N-Term NP_002172.
0.1 mL
Primary
3549