Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Indian Hedgehog/Ihh Rat anti-Mouse, Biotin, Clone: RM0145-3A11, Novus Biologicals
Rat Monoclonal Antibody
Manufacturer: Novus Biologicals NBP122521B
Description
Indian Hedgehog/Ihh Monoclonal antibody specifically detects Indian Hedgehog/Ihh in Mouse samples. It is validated for Western Blot.Specifications
Indian Hedgehog/Ihh | |
RM0145-3A11 | |
Western Blot | |
Purified | |
IHH | |
Mouse recombinant protein of N-Terminal fragment of ihh protein | |
Protein G purified | |
Stem Cell Signaling Pathway | |
Primary | |
3549 | |
Mouse |
Western Blot | |
Biotin | |
PBS with 0.05% Sodium Azide | |
BDA1Indian hedgehog homolog (Drosophila), HHG2, HHG-2, Indian hedgehog, Indian hedgehog (Drosophila) homolog, Indian hedgehog homolog, indian hedgehog protein | |
Rat | |
IgG2 | |
0.1 ml | |
Store at 4C in the dark. | |
Monoclonal | |
This was produced from a hybridoma (mouse myeloma) fused with spleen cells from a rat immunized with mouse recombinant protein of the N-Terminal fragment of ihh protein. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title