Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Indian Hedgehog/Ihh Rat anti-Mouse, Clone: RM0145-3A11, Novus Biologicals™
Rat Monoclonal Antibody
Specifications
Antigen | Indian Hedgehog/Ihh |
---|---|
Clone | RM0145-3A11 |
Concentration | LYOPH |
Dilution | Western Blot 1:500-1:1000 |
Applications | Western Blot |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP122521
|
Novus Biologicals
NBP122521 |
0.1 mg | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
NB018456
|
Novus Biologicals
NBP1225210.025MG |
0.025 mg | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
Indian Hedgehog/Ihh Monoclonal antibody specifically detects Indian Hedgehog/Ihh in Mouse samples. It is validated for Western Blot.Specifications
Indian Hedgehog/Ihh | |
LYOPH | |
Western Blot | |
Unconjugated | |
Rat | |
Stem Cell Signaling Pathway | |
BDA1Indian hedgehog homolog (Drosophila), HHG2, HHG-2, Indian hedgehog, Indian hedgehog (Drosophila) homolog, Indian hedgehog homolog, indian hedgehog protein | |
IHH | |
IgG2 | |
Protein G purified | |
This was produced from a hybridoma (mouse myeloma) fused with spleen cells from a rat immunized with mouse recombinant protein of the N-Terminal fragment of ihh protein. |
RM0145-3A11 | |
Western Blot 1:500-1:1000 | |
Monoclonal | |
Purified | |
RUO | |
Mouse | |
3549 | |
Mouse recombinant protein of N-Terminal fragment of ihh protein | |
Primary | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title