Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Indicator A, Reagents
Click to view available options
Quantity:
1 L
4 L
Packaging:
Glass Bottle
Poly Bottle
Specifications
Specifications
Chemical Name or Material | Indicator A |
CAS | 7732-18-5, 3625-57-8 |
Boiling Point | 100°C |
Synonym | Nile blue sulfate |
Grade | Reagent |
Packaging | Poly Bottle |
Specific Gravity | 1.001 |
Color | Blue |
Quantity | 4 L |
Physical Form | Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction