Learn More
Sklar InstrumentsSupplier Diversity Partner Indicator Tape, Sklar, 1/2 in. x 60 yds, Semi-Crepe Paper, Disposable, Non-Sterile. Sklar's Steam indicator tape is used for visual assurance that the package has been exposed to the sterilizing process. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 101130
Indicator Tape, Sklar, 1/2 in. x 60 yds, Semi-Crepe Paper, Disposable, Non-Sterile. Sklar's Steam indicator tape is used for visual assurance that the package has been exposed to the sterilizing process. The color of the strip changes from baige to black during the process. This tape is 1/2 inch X 60 yards.
Catalog No. 50-143-6928