Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Indium arsenide, 99% (metals basis)

In semiconductor electronics

Manufacturer:  Alfa Aesar™ 2265109

Quantity 10g

 View more versions of this product

Catalog No. AA2265109


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

1303-11-3
189.74
UN1557
Indium arsenide
InAs
MFCD00016144
14,4949

Specifications

1303-11-3
InAs
MFCD00016144
14,4949
5mm & down
10g
99%, (metals basis)
Pieces
189.74
UN1557
Odorless
936°C
Indium arsenide

Safety and Handling

SDS & Safety Documents

GHS H Statement
H301-H331
Toxic if swallowed.
Toxic if inhaled.

GHS P Statement
P261-P301+P310-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ARSENIC COMPOUNDS, SOLID, N.O.S.

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit