Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ Indium foil, 0.05mm (0.002 in.) thick, 99.99% (metals basis)
Click to view available options
Quantity:
25 x 25mm
50 x 50mm
100 x 100mm
Specifications
Quantity | 50 x 50mm |
Weight | ≈0.23g/25x25mm |
Odor | Odorless |
Physical Form | Foil |
Percent Purity | 99.99% |
Chemical Name or Material | Indium foil, 0.05mm (0.002 in.) thick |
Assay | (metals basis) |
Safety and Handling
EINECSNumber : 231-180-0
TSCA : Yes
Recommended Storage : Ambient temperatures