Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Indium foil, 0.25mm (0.01 in.) thick, Puratronic™, 99.9975% (metals basis)

Click to view available options
Quantity:
50 x 50mm
100 x 100mm
7440-74-6
In
114.818
MFCD00134048
APFVFJFRJDLVQX-UHFFFAOYSA-N
indium, elemental,unii-045a6v3vfx,powder,shot,powder,-100 mesh,indio,shot, tear drop, 4mm 0.2in,ingot, polycrystalline, puratronic,shot, 5mm 0.2in & down, puratronic,wire, 0.5mm 0.02in dia, puratronic
5359967
CHEBI:30430
indium
[In]
7440-74-6
In
114.818
MFCD00134048
APFVFJFRJDLVQX-UHFFFAOYSA-N
indium, elemental,unii-045a6v3vfx,powder,shot,powder,-100 mesh,indio,shot, tear drop, 4mm 0.2in,ingot, polycrystalline, puratronic,shot, 5mm 0.2in & down, puratronic,wire, 0.5mm 0.02in dia, puratronic
5359967
CHEBI:30430
indium
[In]
Quantity 100 x 100mm
Weight ≈4.6g/50x50mm
Odor Odorless
Physical Form Foil
Percent Purity 99.9975%
Grade Puratronic™
Chemical Name or Material Indium foil, 0.25mm (0.01 in.) thick
Assay (metals basis)

EINECSNumber : 231-180-0

TSCA : Yes

Recommended Storage : Ambient temperatures