Alfa Aesar™ Indium foil, 0.25mm thick, 99.99% (metals basis), Alfa Aesar™

Manufacturer: Alfa Aesar™ 13982GJ

Quantity 100 x 150mm

 View more versions of this product

Catalog No. AA13982GJ

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-74-6
    MDL Number MFCD00134048
    Chemical Name or Material Indium

    Specifications

    Quantity 100 x 150mm
    Density 7.31g/cm3 at 20°C
    Physical Form Foil, 0.25mm (0.01 in.) thick
    EINECS Number 231-180-0
    Assay Percent Range 99.99% (Metals basis)
    MDL Number MFCD00134048
    CAS 7440-74-6
    Odor Odorless
    Appearance Silvery-white foil
    Chemical Name or Material Indium

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit