Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ Indium(I) chloride, anhydrous, 99.995% (metals basis)
Click to view available options
Quantity:
0.5g
5g
25g
Description
- Packaged under argon
Specifications
UN Number | UN3260 |
Beilstein | EM>13, |
Solubility Information | Soluble in water and THF. |
Formula Weight | 150.27 |
Physical Form | Crystalline |
Percent Purity | 99.995% |
Density | 4.19 g/mL |
Odor | Chlorine-like |
Sensitivity | Moisture sensitive |
Boiling Point | 608°C |
Show More |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
EINECSNumber : 236-693-3
TSCA : Yes
storageNote1 :
- Packaged under argon
Recommended Storage : Ambient temperatures