Alfa Aesar™ Indium(I) chloride, Puratronic™, 99.999% (metals basis)

Manufacturer: Alfa Aesar™ 1070714

Quantity 25g

 View more versions of this product

Catalog No. AA1070714

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13465-10-6
    Formula Weight 150.27
    MDL Number MFCD00148875
    UN Number UN3260
    Chemical Name or Material Indium(I) chloride
    Grade Puratronic™

    Specifications

    Formula Weight 150.27
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00148875
    Odor Chlorine-like
    Physical Form Crystalline Lump
    Sensitivity Moisture Sensitive
    Melting Point ≈225°C
    Quantity 25g
    Health Hazard 1 Danger
    TSCA TSCA
    Boiling Point 608°C
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
    CAS 13465-10-6
    UN Number UN3260
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Density 4.19g/mL
    Chemical Name or Material Indium(I) chloride
    Assay 99.999%, (metals basis)
    Grade Puratronic™

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit