Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Indium ICP Standard, 1mL = 10mg In (10,000ppm In), Ricca Chemical

Click to view available options
Quantity:
50mL
500mL
Packaging:
Poly natural
7440-74-6,7697-37-2,7732-18-5
7440-74-6,7697-37-2,7732-18-5
CAS Min % 64.0,33.56,0.64
CAS Max % 66.0,34.93,0.66
Physical Form Liquid
Packaging Poly natural
Concentration or Composition Notes 10,000ppm In
Equivalence 1 mL = 10 mg In
Color Colorless
Quantity 500mL
Chemical Name or Material Indium ICP Standard