Alfa Aesar™ Indium(III) 2,4-pentanedionate, 98%

Used in catalysts

Manufacturer: Alfa Aesar™ 1379403

Quantity 1g

 View more versions of this product

Catalog No. AA1379403

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 14405-45-9
    Formula Weight 412.15
    MDL Number MFCD00013494
    Synonym Indium(III) acetylacetonate
    Chemical Name or Material Indium(III) 2,4-pentanedionate

    Specifications

    Formula Weight 412.15
    Synonym Indium(III) acetylacetonate
    Physical Form Powder
    Melting Point 186°C (dec.)
    TSCA TSCA
    Solubility Information Soluble in benzene
    Quantity 1g
    MDL Number MFCD00013494
    CAS 14405-45-9
    Chemical Name or Material Indium(III) 2,4-pentanedionate
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit