Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Indium(III) acetate, 99.99%, (trace metal basis), anhydrous, ACROS Organics™

Click to view available options
Quantity:
10g
50g
Packaging:
Glass bottle
25114-58-3
C6H9InO6
291.95
LGHOYKSQIQISBI-UHFFFAOYSA-N
indium acetate,acetic acid, indium 3+ salt
24191761
acetic acid;indium
CC(=O)O.[In]
25114-58-3
C6H9InO6
291.95
LGHOYKSQIQISBI-UHFFFAOYSA-N
indium acetate,acetic acid, indium 3+ salt
24191761
acetic acid;indium
CC(=O)O.[In]
for ultra trace analysis
Assay Percent Range 35.3 to 43.3% (In) (Complexometry)
Solubility Information Solubility in water: soluble
Formula Weight 291.95
Physical Form Crystalline Powder
Percent Purity 99.99%
Grade Trace Metal Basis
Packaging Glass bottle
Solubility Solubility in water: soluble.
Total Trace Metal Impurities 100ppm max.
Color White
Melting Point 270.0°C
Quantity 10g
Chemical Name or Material Indium(III) acetate
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash

GHS Signal Word: Warning