Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Indium(III) bromide, 99.99%, (trace metal basis), Acros Organics

Click to view available options
Quantity:
10g
50g
Packaging:
Glass bottle
13465-09-3
Br3In
354.52
MFCD00011057
13465-09-3
Br3In
354.52
MFCD00011057
for ultra trace analysis
Assay Percent Range 99.99%
Linear Formula InBr3
Merck Index 15, 4997
Formula Weight 354.52
Physical Form Crystalline Powder
Percent Purity 99.99%
Grade Trace Metal Basis
Indium (In) 31.7 to 33.0 %
Packaging Glass bottle
Total Trace Metal Impurities 100ppm max.
Boiling Point 656.0°C
Color White
Melting Point 220.0°C
Quantity 10g
Chemical Name or Material Indium(III) bromide
Show More Show Less

GHS H Statement
Causes skin irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

GHS Signal Word: Danger