Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Indium(III) chloride, 99.995%, (trace metal basis), ACROS Organics™

Click to view available options
Quantity:
10g
50g
250g
Packaging:
Glass bottle
10025-82-8
Cl3In
221.18
10025-82-8
Cl3In
221.18
For ultra trace analysis
Assay Percent Range 99.995%
Linear Formula InCl3
Merck Index 15,4998
Solubility Information Solubility in water: reacts. Other solubilities: soluble in benzene
Formula Weight 221.18
Physical Form Crystalline Powder
Percent Purity 99.995%
Grade Trace Metal Basis
Packaging Glass bottle
Solubility Solubility in water: reacts. Other solubilities: soluble in benzene
Total Trace Metal Impurities 0.005% max
Color White
Melting Point 586.0°C
Quantity 250g
Chemical Name or Material Indium(III) chloride
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

EINECSNumber : 233-043-