Alfa Aesar™ Indium(III) chloride, anhydrous, 99.999% (metals basis)

Manufacturer: Alfa Aesar™ 1185603

Quantity 1g

 View more versions of this product

Catalog No. AA1185603

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 10025-82-8
    Formula Weight 221.18
    MDL Number MFCD00011058
    UN Number UN3260
    Merck Index 14,4958
    Chemical Name or Material Indium(III) chloride, Anhydrous

    Specifications

    Formula Weight 221.18
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Sensitivity Hygroscopic
    Color White
    Health Hazard 2 GHS H Statement
    H314-H318-H302
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.
    Density 3.460g/mL
    Physical Form Crystalline
    MDL Number MFCD00011058
    CAS 10025-82-8
    UN Number UN3260
    Merck Index 14,4958
    Boiling Point 800°C
    Melting Point 586°C
    Quantity 1g
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
    TSCA TSCA
    Odor Odorless
    Health Hazard 1 Danger
    RTECS Number NL1400000
    Chemical Name or Material Indium(III) chloride, Anhydrous
    Assay 99.999%, (metals basis)

    GHS H Statement
    H314-H318-H302
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit