Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Indium(III) chloride hydrate, 99.99% (metals basis)

Manufacturer:  Alfa Aesar™ 1185909

Quantity 10g

 View more versions of this product

Catalog No. AA1185909


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

143983-91-9
MFCD00011058
14,4958
221.18 (Anhydrous)
UN3260
Indium(III) chloride hydrate

Specifications

143983-91-9
221.18 (Anhydrous)
UN3260
Soluble in water
White
Indium(III) chloride hydrate
Crystalline
MFCD00011058
14,4958
Hygroscopic
10g
99.99%, (metals basis)

Safety and Handling

SDS & Safety Documents

GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit