Indium(III) chloride, White to off-white powder, 99.999% (Metals basis), Alfa Aesar™

Manufacturer: Alfa Aesar 4434922

Quantity 100g

 View more versions of this product

Catalog No. AA4434922

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 10025-82-8
    Physical Form Powder
    Assay Percent Range 99.999% (Metals basis)
    Molecular Formula InCl3
    Formula Weight 221.18
    MDL Number MFCD00011058
    Chemical Name or Material Indium(III) chloride

    Specifications

    Physical Form Powder
    Formula Weight 221.18
    Quantity 100g
    Color White to off-white
    EINECS Number 233-043-0
    Assay Percent Range 99.999% (Metals basis)
    Molecular Formula InCl3
    MDL Number MFCD00011058
    CAS 10025-82-8
    Health Hazard 1 H301-H314-H318: Toxic if swallowed. Causes severe skin burns and eye damage. Causes serious eye damage.
    Chemical Name or Material Indium(III) chloride

    H301-H314-H318: Toxic if swallowed. Causes severe skin burns and eye damage. Causes serious eye damage.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit