Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Indium(III) iodide, ultra dry, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog Number p-7052259
Click to view available options
Quantity:
2 g
10 g
50 g
13510-35-5
I3In
495.531
MFCD00016151
RMUKCGUDVKEQPL-UHFFFAOYSA-K
indium triiodide, indium iii iodide, indium iodide ini3, acmc-20akqd, ini3, indium iii iodide, anhydrous, indium iii iodide, anhydrous, powder trace metals basis
83539
triiodoindigane
[In](I)(I)I
13510-35-5
I3In
495.531
MFCD00016151
RMUKCGUDVKEQPL-UHFFFAOYSA-K
indium triiodide, indium iii iodide, indium iodide ini3, acmc-20akqd, ini3, indium iii iodide, anhydrous, indium iii iodide, anhydrous, powder trace metals basis
83539
triiodoindigane
[In](I)(I)I

It is used in the preparation vinyl and methyl indates which undergo regioselective reactions with cinnamyl bromide. It is used as an iodide getter in the Cativa Process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY
Melting Point 210°C
Color Yellow to Brown
Physical Form Beads
Quantity 2 g
UN Number UN3260
Solubility Information Soluble in water.
Formula Weight 495.53
Percent Purity 99.999%
Packaging Ampule
Sensitivity Hygroscopic; Light sensitive
Density 4.69 g/mL
Assay (metals basis)
Chemical Name or Material Indium(III) iodide, Ultra dry
Show More Show Less
Hazard Category H315-H317-H319-H334-H335-H360
Hazard Statement GHS H Statement
H334-H361-H314-H318-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of damaging fertility or the unborn child.
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P261-P264b-P271-P272-P280-P281-P285-P302+P352-P304+P340-P305+P351+P338-P308+P313-P333+P313-P342+P311-P362-P501c
DOTInformation Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
EINECSNumber 236-839-6
TSCA Yes
storageNote1
  • Ampouled under argon
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit